missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, BAKER ANALYZED PHOTREX™, J.T. Baker™

Product Code. 10597301 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10597301 2.5 L 2.5 L
10453861 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10597301 Supplier JT Baker Supplier No. 8706.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Does not interfere with the measurement at the specific wavelength being measured. Cyclohexane, BAKER ANALYZED PHOTREX™, J.T. Baker™ is recommended for use in UV, visible, and IR spectrometry applications.

PHOTREX™ solvents are manufactured to minimize lot-to-lot variability and contaminants that can interfere with UV, and in some cases IR, spectra, including residue after evaporation, and acid and base concentrations. Function testing confirms maximum absorbance in selected wavelengths; 50-100% transmittance windows in IR wavelengths are reported.

  • High quality chemicals for laboratory and specialized industrial use
  • Meets the requirements set forth by the American Chemical Society (ACS)
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.4°C
Absorbance Max. 1.00 at 210nm, Max. 0.50 at 220nm, Max. 0.20 at 230nm, Max. 0.08 at 240nm, Max. 0.03 (at 250nm) - 0.01 (at 300-400nm)
Color Colorless
Density 0.78g/cm3 at 20°C
Quantity 2.5 L
Explosion Limit 1.2 - 8.3 Vol %
Flash Point −18°C
Ignition Point 260°C
Odor Sweet
Packaging Glass
Physical Form Fluid
Residue after Evaporation Max. 0.002%
Linear Formula CH2(CH2)4CH2
Water Max. 0.02%
Boiling Point 81°C
Solubility Information 0.05g/L in water at 20°C
Formula Weight 84.16g/mol
Percent Purity ≥99.0%
Grade Meets ACS specifications
Chemical Name or Material Cyclohexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P331-Do NOT induce vomiting.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.