missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10261051 Shop All J.T.Baker Products
Change view
Click to view available options
:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10261051 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10261051 Supplier JT Baker Supplier No. 8026.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Exceeds ACS Specifications

Specifications

Melting Point 7°C
CAS 110-82-7
Color Max. 10 (APHA)
Density 0.7785 g/cm³ (20°C)
Quantity 1 L
Flash Point -18°C
Residue after Evaporation max. 0.002%
Assay Percent Range min. 99.0%
Molecular Formula C6H12
MDL Number MFCD00003814
Water max. 0.02%
Boiling Point 81°C (1013 hPa)
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES C1CCCCC1
IUPAC Name cyclohexane
Molecular Weight (g/mol) 84.16
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P331-Do NOT induce vomiting.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.