missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, ≥99.8%, Laboratory Reagent, Solstice

Product Code. 15657720
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
4 x 2.5 L
4 x 5 L
150 kg
Packaging:
Aluminum Bottle
Glass Bottle
Metal Can
Metal Drum
Stainless Steel Can
Unit Size:
1 L
10 L
150kg
2.5 L
25 L
5 L
Pack of 4
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15657720 4 x 2.5 L Glass Bottle Pack of 4
15627720 1 L Glass Bottle 1 L
15637720 2.5 L Glass Bottle 2.5 L
15677720 5 L Aluminum Bottle 5 L
15607720 10 L Metal Can 10 L
15667720 4 x 5 L Aluminum Bottle Pack of 4
15647720 25 L Stainless Steel Can 25 L
15617720 150 kg Metal Drum 150kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15657720 Supplier Fluka Supplier No. C1003074X2.5L
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Laboratory Reagent,≥99.8%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.4°C
Color Colorless
Density 0.779g/mL at 25°C
Quantity 4 x 2.5 L
Explosion Limit 0.09
Ignition Point 500°F
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 168.8mmHg (37.7°C), 77mmHg (20°C)
UN Number UN1145
Beilstein 1900225
Boiling Point 81°C (1013 hPa)
Formula Weight 84.16g/mol
Percent Purity ≥99.8%
Chemical Name or Material Cyclohexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.