missing translation for 'onlineSavingsMsg'
Learn More

Solstice Cyclohexane

Product Code. 16605823
Change view
Click to view available options
Quantity:
18 L
45 L
200 L
Unit Size:
18 L
200 L
45 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16605823 200 L 200 L
15617025 18 L 18 L
15975460 45 L 45 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16605823 Supplier Solstice Supplier No. 33117200L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cyclohexane, Puriss. p.a., ACS Reagent, ≥99.5% (GC).

  • ACS general use solvents.
  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis.
  • Suitable for less demanding applications such as routine isocratic HPLC.
  • Variations and decrease issues traceable to solvent contamination.
  • All solvents are tested to meet ACS general use specifications.

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym Hexahydrobenzene
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

pH Not applicable
Melting Point 6.4°C
Color Colorless
CAS Max % ≤100.00
Density 0.78 g/cm3 (20°C)
Quantity 200 L
Flash Point 8.3 %(V)
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line Puriss. p.a.
Linear Formula C6H12
Vapor Pressure 130 hPa (25°C)
Viscosity 0.94 mPa.s (20°C)
UN Number UN1145
Beilstein 1900225
Concentration ≥99.50%
Boiling Point 81°C (1.013 hPa)
Percent Purity ≥99.50%
Grade ACS
Chemical Name or Material Cyclohexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.