missing translation for 'onlineSavingsMsg'
Learn More

Solstice Cyclohexane

Product Code. 16615823
Change view
Click to view available options
Quantity:
7 L
Unit Size:
7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16615823 7 L 7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16615823 Supplier Solstice Supplier No. 33117S7LRC
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cyclohexane, Puriss. p.a., ACS Reagent, ≥99.5% (GC).

  • For HPLC and organic synthesis.

Specifications

pH Not applicable
Melting Point 6.4°C
CAS 110-82-7
Color Colorless
CAS Max % ≤100.00
Density 0.78 g/cm3 (20°C)
Quantity 7 L
Flash Point -18°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line Puriss. p.a.
Molecular Formula C6H12
Linear Formula C6H12
Vapor Pressure 130 hPa (25°C)
MDL Number MFCD00003814
Viscosity 0.94 mPa.s (20°C)
UN Number UN1145
Beilstein 1900225
Concentration ≥99.50%
Synonym Hexahydrobenzene
Boiling Point 81°C (1,013 hPa)
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES C1CCCCC1
IUPAC Name cyclohexane
Molecular Weight (g/mol) 84.16
Percent Purity ≥99.50%
Grade ACS
Chemical Name or Material Cyclohexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.