missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptylacetic acid, 99%

Product Code. 11348326
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348326 1 g 1g
11358326 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348326 Supplier Thermo Scientific Alfa Aesar Supplier No. L15553.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4401-20-1 | C9H16O2 | 156.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4401-20-1
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD01075701
InChI Key DQNWKASPZFJVMJ-UHFFFAOYSA-N
Synonym cycloheptylacetic acid, cycloheptaneacetic acid, 2-cycloheptylaceticacid, pubchem16200, acmc-20ald3
PubChem CID 227401
IUPAC Name 2-cycloheptylacetic acid
SMILES C1CCCC(CC1)CC(=O)O

Specifications

Melting Point 14°C to 15°C
Density 1.023
Boiling Point 110°C to 112°C (1 mmHg)
Refractive Index 1.4725
Quantity 1 g
Beilstein 1935484
Formula Weight 156.23
Percent Purity 99%
Chemical Name or Material Cycloheptylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.