missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptene, 95%, stabilized

Product Code. 10155850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10155850 5 g 5g
10105330 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10155850 Supplier Thermo Scientific Acros Supplier No. 304260050
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 628-92-2 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-92-2
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00004156
InChI Key ZXIJMRYMVAMXQP-UHFFFAOYSA-N
Synonym cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure
PubChem CID 12363
IUPAC Name cycloheptene
SMILES C1CCC=CCC1

Specifications

Melting Point -56°C
Color Colorless
Density 0.8200g/mL
Boiling Point 114.6°C
Flash Point -006°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C7H12
Refractive Index 1.4570 to 1.459
Quantity 5 g
Beilstein 05, 65
Specific Gravity 0.82
Solubility Information Solubility in water: immiscible with water. Other solubilities: soluble in ether,ethanol,benzene
Formula Weight 96.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Cycloheptene, 90%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.