missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptene, 95%, stabilized

Product Code. 10105330 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10105330

Brand: Thermo Scientific Acros 304260250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 628-92-2 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-92-2
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00004156
InChI Key ZXIJMRYMVAMXQP-UHFFFAOYSA-N
Synonym cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure
PubChem CID 12363
IUPAC Name cycloheptene
SMILES C1CCC=CCC1

Specifications

Melting Point -56°C
Color Colorless
Density 0.8200g/mL
Boiling Point 114.6°C
Flash Point -006°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C7H12
Refractive Index 1.4570 to 1.459
Quantity 25 g
Beilstein 05, 65
Specific Gravity 0.82
Solubility Information Solubility in water: immiscible with water. Other solubilities: soluble in ether,ethanol,benzene
Formula Weight 96.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Cycloheptene, 90%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.