missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptanone, 98+%

Product Code. 10793134 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10793134 500 g Glass bottle 500g
10219733 25 g Glass bottle 25g
10776131 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10793134 Supplier Thermo Scientific Acros Supplier No. 111045000
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 502-42-1 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-42-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00004159
InChI Key CGZZMOTZOONQIA-UHFFFAOYSA-N
Synonym suberone, ketocycloheptane, suberon, ketoheptamethylene, cycloheptanon, cycloheptyloxy, chembl18607, cycloheptan-1-one, cyclo-heptanone, 1-oxocycloheptane
PubChem CID 10400
IUPAC Name cycloheptanone
SMILES C1CCCC(=O)CC1

Specifications

Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 179.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C7H12(=O)
Refractive Index 1.4600 to 1.4620
Quantity 500 g
Beilstein 07, 13
Merck Index 15, 2715
Specific Gravity 0.95
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol
Formula Weight 112.17
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Cycloheptanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cycloheptanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.