missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptanone, 98+%

Product Code. 11495453
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495453 250 g 250g
11485453 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495453 Supplier Thermo Scientific Alfa Aesar Supplier No. A13421.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 502-42-1 | C7H12O | 112.172 g/mol

Cycloheptanone is used as a precursor for the synthesis of pharmaceuticals. A spasmolytic agent and vasodilator is produced from this chemical. They are also used in certain stereospecific enzymatic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cycloheptanone is used as a precursor for the synthesis of pharmaceuticals. A spasmolytic agent and vasodilator is produced from this chemical. They are also used in certain stereospecific enzymatic reactions.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents,strong bases, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-42-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00004159
InChI Key CGZZMOTZOONQIA-UHFFFAOYSA-N
Synonym suberone, ketocycloheptane, suberon, ketoheptamethylene, cycloheptanon, cycloheptyloxy, chembl18607, cycloheptan-1-one, cyclo-heptanone, 1-oxocycloheptane
PubChem CID 10400
IUPAC Name cycloheptanone
SMILES C1CCCC(=O)CC1

Specifications

Melting Point -21°C
Density 0.956
Boiling Point 179°C to 181°C
Flash Point 55°C (131°F)
Odor Sweet
Refractive Index 1.461
Quantity 250 g
UN Number UN1224
Beilstein 969823
Merck Index 14,2722
Solubility Information It is insoluble in water.
Formula Weight 112.17
Percent Purity 99%
Chemical Name or Material Cycloheptanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.