missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptanone, 98+%, Thermo Scientific Chemicals

Product Code. 10776131
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-42-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00004159
InChI Key CGZZMOTZOONQIA-UHFFFAOYSA-N
Synonym suberone, ketocycloheptane, suberon, ketoheptamethylene, cycloheptanon, cycloheptyloxy, chembl18607, cycloheptan-1-one, cyclo-heptanone, 1-oxocycloheptane
PubChem CID 10400
IUPAC Name cycloheptanone
SMILES C1CCCC(=O)CC1

Specifications

Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 179.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C7H12(=O)
Refractive Index 1.4600 to 1.4620
Quantity 100 g
Beilstein 07, 13
Merck Index 15, 2715
Specific Gravity 0.95
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol
Formula Weight 112.17
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Cycloheptanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cycloheptanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'