missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptanemethylamine, 98+%

Product Code. 11353636
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353636 1 g 1g
11363636 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353636 Supplier Thermo Scientific Alfa Aesar Supplier No. L14659.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4448-77-5 | C8H17N | 127.231 g/mol

It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix with water.

Notes
Air Sensitive, Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4448-77-5
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
MDL Number MFCD00021674
InChI Key CAOQEOHEZKVYOJ-UHFFFAOYSA-N
Synonym cycloheptanemethylamine, cycloheptylmethylamine, cycloheptanemethanamine, acmc-20agqt, cycloheptylmethyl amine, cycloheptane methylamine, aminomethyl cycloheptane, n-cycloheptylmethyl-amine, aminomethylcycloheptane
PubChem CID 78194
IUPAC Name cycloheptylmethanamine
SMILES C1CCCC(CC1)CN

Specifications

Density 0.884
Boiling Point 98°C to 101°C (35 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.474
Quantity 1 g
UN Number UN2735
Beilstein 2958734
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 127.23
Percent Purity ≥98%
Chemical Name or Material Cycloheptanemethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.