missing translation for 'onlineSavingsMsg'
Learn More

Cyclododecanol, 99%

Product Code. 11455487
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455487 25 g 25g
11465487 100 g 100g
11475487 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455487 Supplier Thermo Scientific Alfa Aesar Supplier No. B23486.14
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 1724-39-6 | C12H24O | 184.323 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1724-39-6
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.323
MDL Number MFCD00003717
InChI Key SFVWPXMPRCIVOK-UHFFFAOYSA-N
Synonym unii-0z645y4jts, dsstox_cid_7429, dsstox_rid_78451, dsstox_gsid_27429, acmc-1bprz
PubChem CID 15595
IUPAC Name cyclododecanol
SMILES C1CCCCCC(CCCCC1)O

Specifications

Melting Point 77°C to 79°C
Flash Point 138°C (280°F)
Quantity 25 g
UN Number UN3077
Formula Weight 184.32
Percent Purity 99%
Chemical Name or Material Cyclododecanol
compliance-icons
Product Identifier
  • Cyclododecanol
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Cyclododecanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.