missing translation for 'onlineSavingsMsg'
Learn More

Cyclododecanol, 98+%

Product Code. 10262792 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10262792 5 g Glass bottle 5g
10488250 100 g Glass bottle 100g
10724221 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10262792 Supplier Thermo Scientific Acros Supplier No. 110960050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1724-39-6 | C12H24O | 184.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1724-39-6
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.32
MDL Number MFCD00003717
InChI Key SFVWPXMPRCIVOK-UHFFFAOYSA-N
Synonym unii-0z645y4jts, dsstox_cid_7429, dsstox_rid_78451, dsstox_gsid_27429, acmc-1bprz
PubChem CID 15595
IUPAC Name cyclododecanol
SMILES C1CCCCCC(CCCCC1)O

Specifications

Melting Point 76.0°C to 79.0°C
Color White
Boiling Point 272.7°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C12H23OH
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents,19.2g/100g benzene,58.5g/100g methanol,33.2g/100g chloroform
Formula Weight 184.32
Percent Purity 98+%
Physical Form Crystalline Solid
Chemical Name or Material Cyclododecanol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Cyclododecanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.