missing translation for 'onlineSavingsMsg'
Learn More

β-Cyclodextrin, MedChemExpress

Product Code. 16401079
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401079 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401079 Supplier Medchem Express Supplier No. HY107201
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress β-Cyclodextrin is a cyclic polysaccharide composed of seven units of glucose (α-D-glucopyranose) linked by α-(1,4) type bonds. β-Cyclodextrin has often been used to enhance the solubility of drugs. β-Cyclodextrin has anti-influenza virus H1N1 activities.

β-Cyclodextrin is a cyclic polysaccharide composed of seven units of glucose (α-D-glucopyranose) linked by α-(1,4) type bonds. β-Cyclodextrin has often been used to enhance the solubility of drugs. β-Cyclodextrin has anti-influenza virus H1N1 activities.

Specifications

Chemical Name or Material β-Cyclodextrin
Physical Form Solid
CAS 7585-39-9
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C42H70O35
Solubility Information H2O : 38.75 mg/mL (34.14 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@]2([H])[C@@H]([C@@H](O)[C@](O[C@]3([H])[C@H](O[C@@](O[C@]4([H])[C@H](O[C@@](O[C@]5([H])[C@H](O[C@@](O[C@@]6([H])[C@H](O)[C@@H](O)[C@@](O[C@@H]6CO)([H])O[C@@]7([H])[C@H](O)[C@@H](O)[C@@](O[C@@H]7CO)([H])O[C@@]8([H])[C@H](O)[C@@H](O)[C@@](O[C@@H]8CO)([H])O2)([H])[C@H](O)[C@H]5O)CO)([H])[C@H](O)[C@H]4O)CO)([H])[C@H](O)[C@H]3O)CO)([H])O1)O
Molecular Weight (g/mol) 1134.98
Formula Weight 1134.98
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.