missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 68168-23-0 |
---|---|
Molecular Formula | C42H70O35 |
Molecular Weight (g/mol) | 1134.99 |
MDL Number | MFCD00150811 |
InChI Key | WHGYBXFWUBPSRW-UHFFFAOYSA-N |
Synonym | beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, puriss hplc |
PubChem CID | 53486154 |
IUPAC Name | 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol |
SMILES | OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10665272
|
Thermo Scientific Acros
227280050 |
5 g | Glass bottle |
N/A
|
N/A | ||||
10308560
|
Thermo Scientific Acros
227280250 |
25 g | Glass bottle |
N/A
|
N/A | ||||
10532251
|
Thermo Scientific Acros
227281000 |
100 g | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentChemical Identifiers
68168-23-0 | |
1134.99 | |
WHGYBXFWUBPSRW-UHFFFAOYSA-N | |
53486154 | |
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O |
C42H70O35 | |
MFCD00150811 | |
beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, puriss hplc | |
5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol |
Specifications
68168-23-0 | |
100.0 | |
White | |
Authentic | |
C42H70O35 | |
06,151; 08,133; 09,129; 12,151; 15,107 | |
+ 161 | |
(1% in water) Clear colorless | |
5ppm toluene max. | |
5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol | |
53486154 | |
99% | |
Crystalline Powder |
98.5 | |
298°C to 300°C | |
5.0 to 8.0 (1% aq. soln.) | |
98.5% min. (ELSD) (HPLC) | |
MFCD00150811 | |
14, 2718 | |
beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, puriss hplc | |
WHGYBXFWUBPSRW-UHFFFAOYSA-N | |
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O | |
1134.99 | |
1135.01 | |
1% max. Alpha-and gamma-cyclodextrin (HPLC) | |
β-Cyclodextrin hydrate, 99% |
Safety and Handling
missing translation for 'tsca' : TSCA
RUO – Research Use Only