missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutylamine hydrochloride, 97%

Product Code. 11461020
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461020 5 g 5g
11471020 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461020 Supplier Thermo Scientific Alfa Aesar Supplier No. B25400.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6291-01-6 | C4H10ClN | 107.58 g/mol

Cyclobutylamine hydrochloride was used in the preparation of cyclobutylarachidonylamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclobutylamine hydrochloride was used in the preparation of cyclobutylarachidonylamide.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool, dry conditions, in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6291-01-6
Molecular Formula C4H10ClN
Molecular Weight (g/mol) 107.58
MDL Number MFCD00034953
InChI Key NFAZOGXQOWEWBM-UHFFFAOYSA-N
Synonym cyclobutanamine hydrochloride, cyclobutylamine hydrochloride, aminocyclobutane hcl, cyclobutanamine, hydrochloride, cyclobutanaminium chloride, cyclobutylamine hcl, acmc-209wdt, aminocyclobutane hydrochloride, cyclobutylamine hydrochloric acid, cyclobutylamine hydrochloride salt
PubChem CID 12618374
IUPAC Name cyclobutanamine;hydrochloride
SMILES C1CC(C1)N.Cl

Specifications

Melting Point 180°C to 183°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 107.58
Percent Purity 97%
Chemical Name or Material Cyclobutylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.