missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutylamine, 98%

Product Code. 10042650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10042650 1 mL Glass bottle 1mL
10667094 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10042650 Supplier Thermo Scientific Acros Supplier No. 209110010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2516-34-9 | C4H9N | 71.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-34-9
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.11
MDL Number MFCD00001328
InChI Key KZZKOVLJUKWSKX-UHFFFAOYSA-N
Synonym cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride
PubChem CID 75645
IUPAC Name cyclobutanamine
SMILES C1CC(C1)N

Specifications

Color Colorless to Yellow
Density 0.8300g/mL
Boiling Point 81.0°C to 82.0°C
Flash Point −4°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C4H7NH2
Refractive Index 1.4366 to 1.4386
Quantity 1 mL
Beilstein 12, 4
Specific Gravity 0.83
Formula Weight 71.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclobutylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclobutylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.