missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutylamine, 98%, Thermo Scientific Chemicals

Catalog Number 10667094
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
2516-34-9
C4H9N
71.11
MFCD00001328
KZZKOVLJUKWSKX-UHFFFAOYSA-N
cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride
75645
cyclobutanamine
C1CC(C1)N
This item is not returnable. View return policy
2516-34-9
C4H9N
71.11
MFCD00001328
KZZKOVLJUKWSKX-UHFFFAOYSA-N
cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride
75645
cyclobutanamine
C1CC(C1)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.8300g/mL
Boiling Point 81.0°C to 82.0°C
Flash Point −4°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C4H7NH2
Refractive Index 1.4366 to 1.4386
Quantity 5 mL
Beilstein 12, 4
Specific Gravity 0.83
Formula Weight 71.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclobutylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclobutylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'