missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanone, 98+%

Product Code. 10030890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10030890 5 g Glass bottle 5g
10560191 1 g Glass bottle 1g
11964831 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10030890 Supplier Thermo Scientific Acros Supplier No. 110930050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1191-95-3 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-95-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00001332
InChI Key SHQSVMDWKBRBGB-UHFFFAOYSA-N
Synonym unii-6pf2sh405u, cyclobutyloxy, cyclobutanone, 98+%, cyclobutanon, cylcobutanone, cylobutanone, 3-cyclobutanone, 1-oxocyclobutane, cyclobutanone, pubchem9021
PubChem CID 14496
IUPAC Name cyclobutanone
SMILES O=C1CCC1

Specifications

Melting Point -50.9°C
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 99.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C4H6(=O)
Refractive Index 1.4205 to 1.4225
Quantity 5 g
Beilstein 07,5
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble
Formula Weight 70.09
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Cyclobutanone
Show More Show Less
compliance-icons
Product Identifier
  • Cyclobutanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.