missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanemethanol, 99%

Product Code. 11457014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457014 25 g 25g
11447014 5 g 5g
11467014 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457014 Supplier Thermo Scientific Alfa Aesar Supplier No. B21480.14
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 4415-82-1 | C5H10O | 86.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4415-82-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00001330
InChI Key WPOPOPFNZYPKAV-UHFFFAOYSA-N
Synonym cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465
PubChem CID 78130
IUPAC Name cyclobutylmethanol
SMILES C1CC(C1)CO

Specifications

Density 0.913
Boiling Point 143°C to 144°C
Flash Point 39°C (102°F)
Refractive Index 1.4465
Quantity 25 g
UN Number UN1105
Beilstein 2036027
Formula Weight 86.13
Percent Purity 99%
Chemical Name or Material Cyclobutanemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.