missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanemethanol, 99%

Product Code. 10347410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347410 25 g Glass bottle 25g
10022360 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347410 Supplier Thermo Scientific Acros Supplier No. 182230250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4415-82-1 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4415-82-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00001330
InChI Key WPOPOPFNZYPKAV-UHFFFAOYSA-N
Synonym cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465
PubChem CID 78130
IUPAC Name cyclobutylmethanol
SMILES C1CC(C1)CO

Specifications

Color Colorless
Density 0.9100g/mL
Boiling Point 143°C to 144°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C4H7CH2OH
Refractive Index 1.4450 to 1.447
Quantity 25 g
Specific Gravity 0.91
Solubility Information Solubility in water: immiscible
Formula Weight 86.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclobutanemethanol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Cyclobutanemethanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.