missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclobutanecarboxylic Acid, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Cyclobutanecarboxylic Acid
Specifications
Specifications
| Chemical Name or Material | Cyclobutanecarboxylic Acid |
| CAS | 3721-95-7 |
| Analyte or Component Names | Cyclobutanecarboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H8 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Cyclobutanecarboxylic Acid, Cyclobutane-1-carboxylic acid, Cyclobutylcarboxylic Acid, NSC 4535 |
| InChI Formula | InChI=1S/C5H8O2/c6-5(7)4-2-1-3-4/h4H,1-3H2,(H,6,7) |
| SMILES | OC(=O)C1CCC1 |
| IUPAC Name | cyclobutanecarboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction