missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarboxylic acid chloride, 99%, Thermo Scientific Chemicals

Product Code. 10487573
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5006-22-4
Molecular Formula C5H7ClO
Molecular Weight (g/mol) 118.56
MDL Number MFCD00001319
InChI Key JFWMYCVMQSLLOO-UHFFFAOYSA-N
Synonym cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1
PubChem CID 78705
IUPAC Name cyclobutanecarbonyl chloride
SMILES C1CC(C1)C(=O)Cl

Specifications

Color Colorless
Density 37g/mL
Boiling Point 145.0°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C4H7COCl
Refractive Index 1.4545 to 1.4565
Quantity 1 g
Beilstein 09, 5
Formula Weight 118.56
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclobutanecarboxylic acid chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclobutanecarboxylic acid chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'