missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarbonyl chloride, 98%

Product Code. 11419836
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419836 10 g 10g
11429836 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419836 Supplier Thermo Scientific Alfa Aesar Supplier No. B20317.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5006-22-4 | C5H7ClO | 118.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5006-22-4
Molecular Formula C5H7ClO
Molecular Weight (g/mol) 118.56
MDL Number MFCD00001319
InChI Key JFWMYCVMQSLLOO-UHFFFAOYSA-N
Synonym cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1
PubChem CID 78705
IUPAC Name cyclobutanecarbonyl chloride
SMILES C1CC(C1)C(=O)Cl

Specifications

Density 1.039
Boiling Point 145°C
Flash Point 37°C (99°F)
Odor Stench
Refractive Index 1.455
Quantity 10 g
UN Number UN2920
Beilstein 956624
Sensitivity Moisture Sensitive
Formula Weight 118.56
Percent Purity 98%
Chemical Name or Material Cyclobutanecarbonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.