missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarbonitrile, 97%

Product Code. 11414063
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414063 25 g 25g
11404063 5 g 5g
11424063 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414063 Supplier Thermo Scientific Alfa Aesar Supplier No. A10309.14
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4426-11-3 | C5H7N | 81.12 g/mol

It is used for research or analysis use only, as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for research or analysis use only, as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4426-11-3
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.12
MDL Number MFCD00013735
InChI Key GFBLFDSCAKHHGX-UHFFFAOYSA-N

Specifications

Density 0.868
Boiling Point 144°C to 146°C
Flash Point 42°C (107°F)
Odor Recognizable
Refractive Index 1.4325
Quantity 25 g
UN Number UN1992
Beilstein 2037573
Synonym cyanocyclobutane, cyclobutyl cyanide, cyclobutancarbonitrile, cyclobutane carbonitrile, acmc-1afpy, cyclobutanecarbonitrile 5g, cyanocyclobutane; cyclobutyl cyanide
Solubility Information Slightly soluble in water.
SMILES C1CC(C1)C#N
IUPAC Name cyclobutanecarbonitrile
PubChem CID 78148
Formula Weight 81.12
Percent Purity 97%
Chemical Name or Material Cyclobutanecarbonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.