missing translation for 'onlineSavingsMsg'
Learn More

Cyclobenzaprine N-Oxide, TRC

Product Code. 18081450
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18081450 100 mg 100mg
18021140 25 mg 25mg
17909649 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18081450 Supplier Toronto Research Chemicals Supplier No. TRCC987760100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6682-26-4
Molecular Formula C20 H21 N O
Molecular Weight (g/mol) 291.39
Synonym 1-Propanamine, 3-(5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-, N-oxide, 5H-Dibenzo[a,d]cycloheptene-δ5,γ-propylamine, N,N-dimethyl-, N-oxide (7CI,8CI), Cyclobenzaprine N-oxide
IUPAC Name 3-(dibenzo[2,1-b:1',2'-e][7]annulen-11-ylidene)-N,N-dimethylpropan-1-amine oxide
SMILES C[N+](C)([O-])CCC=C1c2ccccc2C=Cc3ccccc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H21NO/c1-21(2,22)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20/h3-6,8-14H,7,15H2,1-2H3
Formula Weight 291.1623
Percent Purity >95%
Chemical Name or Material Cyclobenzaprine N-Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.