missing translation for 'onlineSavingsMsg'
Learn More

Cyclobenzaprine hydrochloride, MedChemExpress

Product Code. 16432269
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16432269

Brand: Medchem Express HYB0740

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cyclobenzaprine hydrochloride (MK130 hydrochloride) is a skeletal muscle relaxant and a central nervous system (CNS) depressant.

Cyclobenzaprine hydrochloride (MK130 hydrochloride) is a skeletal muscle relaxant and a central nervous system (CNS) depressant.

Specifications

Chemical Name or Material Cyclobenzaprine hydrochloride
Physical Form Solid
CAS 6202-23-9
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C20H22ClN
For Use With (Application) Neuroscience-Neuromodulation
Synonym MK130 hydrochloride
Solubility Information H2O : ≥ 100 mg/mL (320.67 mM) ∣DMSO : 50 mg/mL (160.33 mM; Need ultrasonic)
SMILES CN(C)CC/C=C1C2=CC=CC=C2C=CC3=CC=CC=C\13.Cl
Molecular Weight (g/mol) 311.85
Formula Weight 311.85
Percent Purity 99.91%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.