missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cyclobenzaprine hydrochloride

Product Code. 17241212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17241212 5 g 5g
17231212 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17241212 Supplier Thermo Scientific Chemicals Supplier No. 466250050
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6202-23-9 | C20H21N·HCl, C20H22ClN | 311.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6202-23-9
Molecular Formula C20H21N·HCl

Specifications

Quantity 5 g
Solubility Solubility aq. soln.: soluble. Other solubilities: soluble in methanol, ethanol
Melting Point 216.0°C to 218.0°C
Formula Weight 311.85
Chemical Name or Material Cyclobenzaprine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.