missing translation for 'onlineSavingsMsg'
Learn More

Cyclobenzaprine-d3 Hydrochloride, TRC

Product Code. 30240512
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240512 25 mg 25mg
30240581 5 mg 5mg
30240777 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240512 Supplier Toronto Research Chemicals Supplier No. TRCC98775225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184983-42-3
Molecular Formula C20 2H3 H18 N . Cl H
Molecular Weight (g/mol) 314.87
Synonym Cyclobenzaprine-D3 hydrochloride, 1-Propanamine, 3-(5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-N-(methyl-d3)-, hydrochloride, 3-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-N-(methyl-d3)-1-propanamine hydrochloride, Cyclobenzaprine-D3 hydrochloride (methyl D3)
IUPAC Name 3-(dibenzo[2,1-b:1',2'-e][7]annulen-11-ylidene)-N-methyl-N-(trideuteriomethyl)propan-1-amine;hydrochloride
SMILES Cl.[2H]C([2H])([2H])N(C)CCC=C1c2ccccc2C=Cc3ccccc13

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H21N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-14H,7,15H2,1-2H3;1H/i1D3;
Formula Weight 314.1629
Chemical Name or Material Cyclobenzaprine-D3 HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.