missing translation for 'onlineSavingsMsg'
Learn More

Cyclo(RGDyK) trifluoroacetate, MedChemExpress

Product Code. 16494376
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494376 100 mg 100mg
16493078 10 mg 10mg
16435817 5 mg 5mg
16419646 50 mg 50mg
16412616 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494376 Supplier Medchem Express Supplier No. HY100563
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cyclo(RGDyK) trifluoroacetate is a potent and selective αVβ3 integrin inhibitor with an IC50 of 20 nM.

Cyclo(RGDyK) trifluoroacetate is a potent and selective αVβ3 integrin inhibitor with an IC50 of 20 nM.

Specifications

Chemical Name or Material Cyclo(RGDyK) trifluoroacetate
CAS 250612-42-1
Color White
Purity Grade Notes Research
Molecular Formula C31H43F6N9O12
Quantity 100 mg
Solubility Information DMSO : ≥ 130 mg/mL (153.35 mM)
SMILES O=C(N[C@H](C(N[C@H]1CCCNC(N)=N)=O)CCCCN)[C@H](NC([C@@H](NC(CNC1=O)=O)CC(O)=O)=O)CC2=CC=C(O)C=C2.O=C(O)C(F)(F)F.O=C(O)C(F)(F)F
Molecular Weight (g/mol) 847.72
Formula Weight 847.72
Percent Purity 99.92%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.