missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Cyclo(Phe-Pro)

Product Code. 16492268
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492268 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492268 Supplier MedChemExpress Supplier No. HYP1934

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cyclo(Phe-Pro) (Cyclo(phenylalanylprolyl)), a Vibrio vulnificus quorum-sensing molecule, inhibits retinoic acid-inducible gene-I (RIG-I) polyubiquitination, through its specific interaction with RIG-I, to blunt IRF-3 activation and type-I IFN production. Cyclo(Phe-Pro) (Cyclo(phenylalanylprolyl)) enhances susceptibility to hepatitis C virus (HCV), as well as Sendai and influenza viruses.

Specifications

Purity 98.06%
CAS 14705-60-3
Content And Storage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Neuroscience-Neuromodulation
Molecular Weight (g/mol) 244.29 g/mol
Product Type Peptides
Quantity 5 mg
Shipping Condition Room temperature
Solubility DMSO : 125 mg/mL (511.69 mM∣Need ultrasonic)
Synonym Cyclo(phenylalanylprolyl)∣ A-64863
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.