missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cyclizine hydrochloride, 98%

Product Code. 15547719 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15547719

Brand: Thermo Scientific Chemicals 461040010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 303-25-3 | C18H22N2·HCl | 302.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 303-25-3
Melting Point 291.0°C to 293.0°C
Loss on Drying 1% max. (120°C, 3 h)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C18H22N2·HCl
Quantity 1 g
Synonym cyclizine hydrochloride, cyclizine hcl, dihydrochlorid, marzine, reis-fit, unii-w0o1nhp4we, marezine hydrochloride, cyclizine monohydrochloride, w0o1nhp4we, 1-diphenylmethyl-4-methylpiperazine monohydrochloride
InChI Key UKPBEPCQTDRZSE-UHFFFAOYSA-N
SMILES CN1CCN(CC1)C(C2=CC=CC=C2)C3=CC=CC=C3.Cl
IUPAC Name 1-benzhydryl-4-methylpiperazine;hydrochloride
Molecular Weight (g/mol) 302.84
PubChem CID 6420019
ChEBI CHEBI:51045
Formula Weight 302.84
Percent Purity 98%
Chemical Name or Material Cyclizine hydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclizine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361-Suspected of damaging fertility or the unborn child.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.