missing translation for 'onlineSavingsMsg'
Learn More

Cyclizine-d3 (N-methyl-d3), CDN

Product Code. 30320086
Change view
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Product Code. Quantity unitSize
30320086 0.05 g 0.05g
30312489 0.01 g 0.01g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30320086

Brand: CDN CDND74120.05G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclizine-d3 (N-methyl-d3)
CAS 1170155-67-5
Molecular Formula C18 D3 H19 N2
Quantity 0.05 g
Synonym Piperazine, 1-(diphenylmethyl)-4-(methyl-d3)- (ACI), 1-(Diphenylmethyl)-4-(methyl-d3)piperazine (ACI), Cyclizine-d3, Cyclizine-D3 (N-methyl-D3), N-Benzhydryl-N'-(methyl-d3)piperazine
InChI Formula InChI=1 S/C18H22N2/c1-19-12-14-20(15-13-19)18(16-8-4-2-5-9-16)17-10-6-3-7-11-17/h2-11,18 H,12-15H2,1H3/i1D3
SMILES [2 H]C([2 H])([2 H])N1CCN(CC1)C(c2ccccc2)c3ccccc3
IUPAC Name 1-benzhydryl-4-(trideuteriomethyl)piperazine
Molecular Weight (g/mol) 269.3992
Formula Weight 269.1971 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.