missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin Y Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557598
This item is not returnable.
View return policy
Description
Immunogen sequence: GNTTSCCVSS SPKLRRNAHS RLESYRPDTD LSREDTGCNL QHISDRENID DLNMEFNPS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.
Specifications
| Cyclin Y | |
| Polyclonal | |
| Unconjugated | |
| CCNY | |
| 1700025H17Rik; 3110050L10Rik; 4631402G10Rik; 5730405I09Rik; C10orf9; CBCP1; CCNX; Ccny; CFP1; Cyclin box protein 1; cyclin fold protein 1; cyclin Y; cyclin-box carrying protein 1; cyclin-X; Cyclin-Y; cyc-Y; RGD1565969; si:dkey-18n12.2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 219771 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8ND76 | |
| CCNY | |
| Recombinant protein corresponding to Human Cyclin Y. Recombinant protein control fragment (Product #RP-97061). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction