missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin Y Polyclonal Antibody

Description
Cyclin Y Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.
Specifications
Specifications
| Antigen | Cyclin Y |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | CCNY |
| Gene Accession No. | Q8ND76 |
| Gene Alias | 1700025H17Rik; 3110050L10Rik; 4631402G10Rik; 5730405I09Rik; C10orf9; CBCP1; CCNX; Ccny; CFP1; Cyclin box protein 1; cyclin fold protein 1; cyclin Y; cyclin-box carrying protein 1; cyclin-X; Cyclin-Y; cyc-Y; RGD1565969; si:dkey-18n12.2 |
| Gene Symbols | CCNY |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?