missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin T2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£213.00 - £459.00
Specifications
| Antigen | Cyclin T2 |
|---|---|
| Dilution | Western Blot WB reported in scientific literature (PMID: 30209133)., Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18441282
|
Novus Biologicals
NBP1-87592-25ul |
25 μL |
£213.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18288826
|
Novus Biologicals
NBP1-87592 |
0.1 mL |
£459.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin T2 Polyclonal specifically detects Cyclin T2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| Cyclin T2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
| Unconjugated | |
| RUO | |
| Human | |
| cyclin T2, cyclin T2a, cyclin T2b, cyclin-T2, CYCT2, FLJ13583, FLJ90560, MGC134840, SDS-stable vimentin-bound DNA fragment HEF42VIM22, subunit of positive elongation transcription factor b | |
| CCNT2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot WB reported in scientific literature (PMID: 30209133)., Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockout Validated | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 905 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KAGSSKHHGPISTTPGIIPQKMSLDKYREKRKLETLDLDVRDHYIAAQVEQQHKQGQSQAASSSSVTSPIKMKIPIANTEKYMADKKEKSGSLKLRIPIPPTDKSASKEELKMKIKVSSSERHSSSDEGSGKSKHSSPH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title