missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cyclin T1 Recombinant Rabbit Monoclonal Antibody (K01_1Q05)
GREENER_CHOICE

Product Code. 30827891
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827891 100 μL
30825802 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827891 Supplier Invitrogen™ Supplier No. MA563808

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Cyclin T1 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a member of the highly conserved cyclin C subfamily. The encoded protein tightly associates with cyclin-dependent kinase 9, and is a major subunit of positive transcription elongation factor b (p-TEFb). In humans, there are multiple forms of positive transcription elongation factor b, which may include one of several different cyclins along with cyclin-dependent kinase 9. The complex containing the encoded cyclin and cyclin-dependent kinase 9 acts as a cofactor of human immunodeficiency virus type 1 (HIV-1) Tat protein, and is both necessary and sufficient for full activation of viral transcription. This cyclin and its kinase partner are also involved in triggering transcript elongation through phosphorylation of the carboxy-terminal domain of the largest RNA polymerase II subunit. Overexpression of this gene is implicated in tumor growth. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin T1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Q05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene Ccnt1
Gene Accession No. O60563, Q9QWV9
Gene Alias 2810478G24Rik; AI115585; CCNT; CCNT1; CDK9-associated C-type protein; cyclin C-related protein; cyclin T1; cyclin T1b; cyclin-T; cyclin-T1; CycT1; HIVE1; human immunodeficiency virus type 1 (HIV-1) expression (elevated) 1
Gene Symbols Ccnt1
Host Species Rabbit
Immunogen Recombinant protein of human Cyclin T1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12455, 315291, 904
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.