missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin T1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577892
This item is not returnable.
View return policy
Description
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (86%), Rat (87%), Chimpanzee (100%), Bovine (84%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the highly conserved cyclin C subfamily. The encoded protein tightly associates with cyclin-dependent kinase 9, and is a major subunit of positive transcription elongation factor b (p-TEFb). In humans, there are multiple forms of positive transcription elongation factor b, which may include one of several different cyclins along with cyclin-dependent kinase 9. The complex containing the encoded cyclin and cyclin-dependent kinase 9 acts as a cofactor of human immunodeficiency virus type 1 (HIV-1) Tat protein, and is both necessary and sufficient for full activation of viral transcription. This cyclin and its kinase partner are also involved in triggering transcript elongation through phosphorylation of the carboxy-terminal domain of the largest RNA polymerase II subunit. Overexpression of this gene is implicated in tumor growth. Alternative splicing results in multiple transcript variants.
Specifications
| Cyclin T1 | |
| Polyclonal | |
| Unconjugated | |
| Ccnt1 | |
| 2810478G24Rik; AI115585; CCNT; CCNT1; CDK9-associated C-type protein; cyclin C-related protein; cyclin T1; cyclin T1b; cyclin-T; cyclin-T1; CycT1; HIVE1; human immunodeficiency virus type 1 (HIV-1) expression (elevated) 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 315291, 904 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.0 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O60563 | |
| Ccnt1 | |
| Recombinant protein encompassing a sequence within the center region of human Cyclin T1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction