missing translation for 'onlineSavingsMsg'
Learn More

Cyclin T1 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18423647 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423647 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423647 Supplier Novus Biologicals Supplier No. NBP281898DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cyclin T1 Polyclonal antibody specifically detects Cyclin T1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin T1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias CCNT, CCNT1, CCNTcyclin-T, CDK9-associated C-type protein, cyclin C-related protein, cyclin T1, cyclin T1b, Cyclin-T, cyclin-T1, CYCT1, HIVE1, human immunodeficiency virus type 1 (HIV-1) expression (elevated) 1, Human immunodeficiency virus-1 expression, subunit of positive elongation transcription factor b
Host Species Rabbit
Immunogen Rabbit polyclonal Cyclin T1 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human Cyclin T1. The immunogen is located within the last 50 amino acids of Cyclin T1. Amino Acid Squence: NTDKKPRPPPLPSEPPPPL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 904
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.