missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin M3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97251-100ul
This item is not returnable.
View return policy
Description
Cyclin M3 Polyclonal antibody specifically detects Cyclin M3 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Cyclin M3 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| ACDP3cyclin-M3, ancient conserved domain protein 3, Ancient conserved domain-containing protein 3, cyclin M3, Cyclin-M3, DKFZp434I1016, FLJ20018, metal transporter CNNM3 | |
| Produced in rabbits immunized with E. coli-derived Human Cyclin M3 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 26505 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction