missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562738
This item is not returnable.
View return policy
Description
Immunogen sequence: DRLYSGVLIT LENCLLPDDK LRFTPSMSSG LDTDTETDLR VVGCEL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
CCNL2 belongs to the cyclin family, Cyclin L subfamily. CCNL2 is a transcriptional regulator which participates in regulating the pre-mRNA splicing process. CCNL2 also modulates the expression of critical apoptotic factor, leading to cell apoptosis.
Specifications
| Cyclin L2 | |
| Polyclonal | |
| Unconjugated | |
| CCNL2 | |
| 1700010A01Rik; 1810019L15Rik; 2010319M22Rik; AA409936; Ania6b; ANIA-6B; AW261738; Ccnl2; CCNM; CCNS; Cyclin Ania-6b; cyclin L2; cyclin M; cyclin S; cyclin-L2; HCLA-150; HCLA-ISO; HLA-ISO; hypothetical protein LOC538796; MNCb-5160; paneth cell enhanced expression; paneth cell-enhanced expression protein; PCEE; SB138 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81669 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96S94 | |
| CCNL2 | |
| Recombinant protein corresponding to Human Cyclin L2. Recombinant protein control fragment (Product #RP-100477). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction