missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin L2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | Cyclin L2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Cyclin L2 Polyclonal antibody specifically detects Cyclin L2 in Human samples. It is validated for Western BlotSpecifications
| Cyclin L2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ania-6b, CCNM, cyclin L2, cyclin M, cyclin-L2, DKFZp761A1210, DKFZp762O195, HCLA-ISO, HLA-ISO, Paneth cell-enhanced expression protein, PCEE, SB138 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-150 of human Cyclin L2 (NP_001307082.1). QLYARKKVDLTHLEGEVEKRKHAIEEAKAQARGLLPGGTQVLDGTSGFSPAPKLVESPKEGKGSKPSPLSVKNTKRRLEGAKKAKADSPVN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 81669 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title