missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin I Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567680
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin shows the highest similarity with cyclin G. The transcript of this gene was found to be expressed constantly during cell cycle progression. The function of this cyclin has not yet been determined.
Specifications
| Cyclin I | |
| Polyclonal | |
| Unconjugated | |
| CCNI | |
| CCNI; CCNI1; CYC1; cyclin I; cyclin ITI; cyclin-I; Cyclin-I-like protein; CYI | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10983, 12453 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14094, Q9Z2V9 | |
| CCNI | |
| Synthesized peptide derived from human Cyclin I. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction