missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin I Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16051
This item is not returnable.
View return policy
Description
Cyclin I Polyclonal specifically detects Cyclin I in Human samples. It is validated for Western Blot.
Specifications
| Cyclin I | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
| CCNI | |
| Recombinant protein encompassing a sequence within the C-terminus region of human cyclin I. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| CYC1, cyclin I, cyclin ITI, cyclin-I, CYI | |
| Rabbit | |
| 43 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 10983 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction