missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin H Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517586
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other family members at physiological levels.
The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin forms a complex with CDK7 kinase and ring finger protein MAT1. The kinase complex is able to phosphorylate CDK2 and CDC2 kinases, thus functions as a CDK-activating kinase (CAK). This cyclin and its kinase partner are components of TFIIH, as well as RNA polymerase II protein complexes. They participate in two different transcriptional regulation processes, suggesting an important link between basal transcription control and the cell cycle machinery.
Specifications
| Cyclin H | |
| Polyclonal | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P51946, Q61458, Q9R1A0 | |
| CCNH | |
| Synthetic peptide corresponding to residues near the carboxy-terminus of cyclin H. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| CCNH | |
| 6330408H09Rik; AI661354; AV102684; AW538719; CAK; CAK complex subunit; Ccnh; CDK-activating kinase complex subunit; CycH; cyclin H; cyclin-dependent kinase-activating kinase complex subunit; cyclin-H; MO15-associated protein; OTTHUMP00000222395; OTTHUMP00000222396; p34; p37 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 66671, 84389, 902 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction