missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin H Antibody (3G6), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59440AF750
This item is not returnable.
View return policy
Description
Cyclin H Monoclonal antibody specifically detects Cyclin H in Human samples. It is validated for Western Blot, ELISA
Specifications
| Cyclin H | |
| Monoclonal | |
| Alexa Fluor 750 | |
| CAK, cyclin H, cyclin-dependent kinase-activating kinase complex subunit, cyclin-H, MO15-associated protein, p34CAK complex subunit, p37CDK-activating kinase complex subunit | |
| Recombinant human Cyclin H (1-323aa) purified from E.coli | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair | |
| 902 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA | |
| 3G6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction