missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin G2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA557013
This item is not returnable.
View return policy
Description
Immunogen sequence: PSVLALCLLN LEVETLKSVE LLEILLLVKK HSKINDTEFF YWRELVSKCL AEYSSPECCK PDLKKLVWIV SRRTAQNLHN SYYSVPELPT IPEGGCFDES ES Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
Cyclin G2 may play a role in growth regulation and in negative regulation of cell cycle progression.
Specifications
| Cyclin G2 | |
| Polyclonal | |
| Unconjugated | |
| CCNG2 | |
| Cyclin G2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 901 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16589 | |
| CCNG2 | |
| Recombit protein corresponding to Human Cyclin G2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction