missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin G Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10897-1-AP-20UL
This item is not returnable.
View return policy
Description
The eukaryotic cell cycle is governed by cyclin-dependent protein kinases (CDKs) whose activities are regulated by cyclins and CDK inhibitors. The protein encoded by this gene is a member of the cyclin family and contains the cyclin box. The encoded protein lacks the protein destabilizing (PEST) sequence that is present in other family members. Transcriptional activation of this gene can be induced by tumor protein p53. Two transcript variants encoding the same protein have been identified for this gene.Specifications
| Cyclin G | |
| Polyclonal | |
| Unconjugated | |
| CCNG1 | |
| CCNG, CCNG1, CYCG1, Cyclin G, cyclin G1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12450, 25405, 900 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P39950, P51945, P51959 | |
| CCNG1 | |
| Cyclin G Fusion Protein Ag1157 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction