missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin E2 Recombinant Rabbit Monoclonal Antibody (24GB4750)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | Cyclin E2 |
|---|---|
| Clone | 24GB4750 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276064
![]() |
Invitrogen™
MA557373 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274838
|
Invitrogen™
MA557374 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin E2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
This gene encodes WW domain-containing binding protein 4. The WW domain represents a small and compact globular structure that interacts with proline-rich ligands. This encoded protein is a general spliceosomal protein that may play a role in cross-intron bridging of U1 and U2 snRNPs in the spliceosomal complex A.Specifications
| Cyclin E2 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| CCNE2 | |
| Ccne2; CYCE2; cyclin E2; G1/S-specific cyclin-E2 | |
| CCNE2 | |
| Primary | |
| Affinity chromatography |
| 24GB4750 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O96020 | |
| 9134 | |
| A synthesized peptide derived from human Cyclin E2 (aa 115-129). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title